Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033324.2 | NP_201581.2 | 2 | 400 | ||
GenBank mRNA | AB086857.1 | - | 2 | 400 | ||
AF201683.1 | - | 1 | 54 | |||
AK048056.1 | - | 1 | 201 | |||
AK086233.1 | - | 2 | 557 | |||
BC051013.1 | - | 1 | 173 | |||
BC062919.1 | - | 2 | 420 | 6833280 | ||
BC066118.1 | - | 2 | 447 | 6400561 |