siRNA Details
Gene Symbol: Slc9b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178877.6 | NP_849208.4 | 3, 4 | 412 | ||
GenBank mRNA | AK076505.1 | - | 4, 5 | 453 | ||
BC090977.1 | - | 3, 4 | 365 | 30503721 |

Gene Symbol: Slc9b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178877.6 | NP_849208.4 | 3, 4 | 412 | ||
GenBank mRNA | AK076505.1 | - | 4, 5 | 453 | ||
BC090977.1 | - | 3, 4 | 365 | 30503721 |