Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178640.2 | NP_848755.1 | 9 | 1216 | ||
GenBank mRNA | AB116655.1 | - | 9 | 1085 | ||
AK035259.1 | - | 9 | 1217 | |||
AK041022.1 | - | 9 | 1212 | |||
AK084275.1 | - | 9 | 1216 | |||
AK085691.1 | - | 9 | 1218 | |||
AK134868.1 | - | 8 | 979 | |||
AK144880.1 | - | 9 | 1047 | |||
AK151677.1 | - | 9 | 1266 | |||
AK153362.1 | - | 9 | 1266 | |||
AK167635.1 | - | 9 | 1217 | |||
BC085110.1 | - | 9 | 1225 | 30735904 |