Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173865.3 | NP_776290.1 | 2 | 1282 | ||
GenBank mRNA | AK089802.1 | - | 2 | 1283 | ||
AK154819.1 | - | 2 | 1255 | |||
AK192565.1 | - | 1 | 24 | |||
BC096596.1 | - | 2 | 1318 | 4239523 | ||
BC116280.1 | - | 2 | 1090 | 40094408 | ||
BC116281.1 | - | 2 | 1090 | 40094411 |