Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3gap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163754.1 | NP_001157226.1 | 28, 29 | 3298 | ||
GenBank mRNA | AK030955.1 | - | 18, 19 | 2298 | ||
AK031191.1 | - | 18, 19 | 2207 | |||
AK048759.1 | - | 5, 6 | 440 | |||
AK122385.1 | - | 15, 16 | 1866 | |||
AK149265.1 | - | 5, 6 | 469 | |||
BC035944.1 | - | 19, 20 | 2288 | |||
BC052505.1 | - | 3, 4 | 396 | |||
BC057872.1 | - | 19, 20 | 2316 | 5294131 |