Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195390.1 | NP_001182319.1 | 4 | 408 | ||
NM_133850.2 | NP_598611.2 | 3 | 333 | |||
GenBank mRNA | AK008447.1 | - | 3 | 288 | ||
AK009274.1 | - | 3 | 308 | |||
AK046582.1 | - | 3 | 283 | |||
AK077349.1 | - | 3 | 274 | |||
AK078774.1 | - | 3 | 292 | |||
AK164805.1 | - | 3 | 242 | |||
AK169037.1 | - | 4 | 355 | |||
BC003310.1 | - | 3 | 266 | |||
BC065794.1 | - | 3 | 234 | |||
BC092215.1 | - | 3 | 250 | 6819383 | ||
BC094216.1 | - | 3 | 290 | 6820900 | ||
BY295213.1 | - | 3 | 334 |