Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133859.2 | NP_598620.2 | 3 | 874 | ||
GenBank mRNA | AK002518.1 | - | 3 | 837 | ||
AK076084.1 | - | 3 | 840 | |||
AK131879.1 | - | 838 | ||||
AK143728.1 | - | 1426 | ||||
BC005485.1 | - | 3 | 821 | 3485412 |