Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK147106.1 | - | 1 | 693 | ||
AK162481.1 | - | 1 | 401 | |||
AK220375.1 | - | 1 | 690 | |||
BC145902.1 | - | 1 | 372 | 40131205 |