Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133872.2 | NP_598633.2 | 11 | 1523 | ||
GenBank mRNA | AK129170.1 | - | 11 | 1463 | ||
BC059885.1 | - | 11 | 1510 | 6815975 |