Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081557.3 | NP_001075026.1 | 3 | 278 | ||
NM_001195565.1 | NP_001182494.1 | 3 | 278 | |||
GenBank mRNA | AK009988.1 | - | 2 | 178 | ||
AK034038.1 | - | 3 | 215 | |||
AK076589.1 | - | 2 | 91 | |||
AK144150.1 | - | 3 | 248 | |||
AK158710.1 | - | 3 | 279 | |||
BC150740.1 | - | 2 | 127 | 9087651 |