Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133879.2 | NP_598640.2 | 7 | 1391 | ||
GenBank mRNA | AK027912.1 | - | 6 | 1875 | ||
AM161454.1 | - | 7 | 1339 | |||
BC005515.1 | - | 6 | 1500 | 3493611 | ||
CX202255.1 | - | 5 | 568 |