Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133891.3 | NP_598652.3 | 14 | 2116 | ||
GenBank mRNA | AK014708.1 | - | 1 | 369 | ||
AY249865.1 | - | 14 | 1872 | |||
AY249866.1 | - | 14 | 1845 | |||
BC010258.1 | - | 2 | 310 | |||
BC025941.1 | - | 5 | 678 | |||
BC113167.1 | - | 14 | 1872 | 40059854 | ||
BC113169.1 | - | 14 | 1872 | 40059860 |