Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133904.2 | NP_598665.2 | 51, 52 | 7222 | ||
GenBank mRNA | AK076301.1 | - | 10, 11 | 1312 | ||
AK147463.1 | - | 40, 41 | 5417 | |||
AY451394.1 | - | 51, 52 | 7222 | |||
BC022940.1 | - | 17, 18 | 2308 |