Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emilin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133918.2 | NP_598679.1 | 8 | 2949 | ||
GenBank mRNA | AK029337.1 | - | 8 | 2920 | ||
AK052368.1 | - | 8 | 3029 | |||
AK088995.1 | - | 8 | 3015 | |||
AK140388.1 | - | 6 | 2365 | |||
AK155486.1 | - | 8 | 3251 | |||
BC005481.1 | - | 8 | 2949 | 3485057 |