Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053194.4 | NP_444424.1 | 5 | 989 | ||
GenBank mRNA | AF288813.1 | - | 5 | 988 | ||
AK031466.1 | - | 5 | 936 | |||
AK080192.1 | - | 5 | 932 | |||
AK155282.1 | - | 5 | 964 | |||
AK165536.1 | - | 5 | 966 | |||
AK167746.1 | - | 5 | 992 | |||
AK169367.1 | - | 5 | 953 | |||
AK171093.1 | - | 1 | 2055 | |||
BC022917.1 | - | 5 | 966 | 4460521 |