Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005863.2 | NP_001005863.1 | 9 | 3374 | ||
NM_001005864.3 | NP_001005864.1 | 7 | 980 | |||
NM_001005865.3 | NP_001005865.2 | 5 | 993 | |||
NM_001286413.1 | NP_001273342.1 | 9 | 3256 | |||
GenBank mRNA | AF173380.1 | - | 5 | 832 | ||
AF493235.1 | - | 5 | 897 | |||
AK030510.1 | - | 5 | 957 | |||
AK031693.1 | - | 5 | 927 | |||
AK035576.1 | - | 3 | 361 | |||
AK129324.1 | - | 4 | 2425 | |||
AY246699.1 | - | 5 | 655 | |||
AY626781.1 | - | 5 | 990 | |||
AY626782.1 | - | 7 | 978 | |||
AY626783.1 | - | 8 | 3196 | |||
BC041777.1 | - | 8 | 2039 | 4240274 | ||
BC042206.1 | - | 8 | 2039 | 4240274 | ||
BC043321.1 | - | 7 | 944 | |||
BC089009.1 | - | 9 | 3256 | 30543240 |