Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACVR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004302.4 | NP_004293.1 | 4 | 699 | ||
NM_020327.3 | NP_064732.3 | 4 | 651 | |||
NM_020328.3 | NP_064733.3 | 4 | 699 | |||
XM_011538966.2 | XP_011537268.1 | 4 | 677 | |||
XM_011538967.2 | XP_011537269.1 | 4 | 677 | |||
XM_017020199.1 | XP_016875688.1 | 4 | 1397 | |||
XM_017020200.1 | XP_016875689.1 | 5 | 792 | |||
XM_017020201.1 | XP_016875690.1 | 4 | 677 | |||
GenBank mRNA | AK299120.1 | - | 4 | 662 | ||
AK299496.1 | - | 4 | 651 | |||
BC000254.1 | - | 4 | 648 | 3352720 | ||
BC040531.1 | - | 4 | 696 | 5260767 | ||
BT007072.1 | - | 4 | 622 | |||
L10125.1 | - | 4 | 661 | |||
L10126.1 | - | 4 | 622 | |||
U14722.1 | - | 4 | 622 | |||
Z22536.1 | - | 4 | 622 |