Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033209.3 | NP_001028381.1 | 5 | 628 | ||
NM_001199568.1 | NP_001186497.1 | 4 | 558 | |||
GenBank mRNA | AK143476.1 | - | 5 | 628 | ||
AK164186.1 | - | 4 | 352 | |||
AK165440.1 | - | 5 | 425 | |||
AK165518.1 | - | 5 | 425 | |||
AK165759.1 | - | 4 | 354 | |||
AK170197.1 | - | 4 | 371 | |||
BC080743.1 | - | 5 | 402 | |||
BC138244.1 | - | 5 | 577 | 8861263 |