Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153133.2 | NP_694773.1 | 4 | 1927 | ||
GenBank mRNA | AB081322.1 | - | 4 | 1866 | ||
AF372838.1 | - | 4 | 1927 | |||
AK132181.1 | - | 1998 | ||||
BC141318.1 | - | 4 | 1906 | 9055930 |