Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002536.2 | NP_002527.1 | 6 | 1281 | ||
GenBank mRNA | AB449912.1 | - | 6 | 1170 | ||
AK293521.1 | - | 5 | 1111 | |||
AK293875.1 | - | 6 | 1240 | |||
AK294776.1 | - | 6 | 1179 | |||
AK297924.1 | - | 3 | 686 | |||
AK299561.1 | - | 1 | 209 | |||
AK316199.1 | - | 4 | 961 | |||
BC026050.1 | - | 1 | 204 | |||
BC036942.1 | - | 3 | 710 | |||
BC052609.1 | - | 5 | 1071 | |||
BC082255.1 | - | 4 | 1181 | |||
BC101817.1 | - | 6 | 1207 | 8069323 | ||
BC101819.1 | - | 6 | 1207 | 8069325 | ||
BC125088.1 | - | 6 | 1196 | 40117866 | ||
BC125089.1 | - | 5 | 1041 | |||
L08240.1 | - | 5 | 1060 |