Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175331.3 | NP_780540.2 | 8 | 995 | ||
GenBank mRNA | AK147488.1 | - | 8 | 964 | ||
AK172248.1 | - | 8 | 952 | |||
AY651022.1 | - | 714 |