Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134010.2 | NP_598771.1 | 13 | 1192 | ||
GenBank mRNA | AK088069.1 | - | 13 | 1193 | ||
AK088303.1 | - | 13 | 1169 | |||
AK088625.1 | - | 13 | 1162 | |||
AK133938.1 | - | 13 | 1171 | |||
AK161665.1 | - | 13 | 1180 | |||
AK199190.1 | - | 3 | 147 | |||
BC004655.1 | - | 9 | 667 | |||
BC057591.1 | - | 13 | 1159 | 6402677 | ||
BC092261.1 | - | 13 | 1169 | 2646396 |