Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002764.1 | NP_001002764.1 | 3 | 1923 | ||
GenBank mRNA | AK129203.1 | - | 2 | 542 | ||
BC037179.1 | - | 2 | 275 | |||
BC047279.1 | - | 3 | 1935 | |||
BC066040.1 | - | 3 | 1923 | 6410245 | ||
BC077715.1 | - | 3 | 1886 |