Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178909.4 | NP_849240.1 | 4, 5 | 597 | ||
GenBank mRNA | AK033294.1 | - | 4, 5 | 543 | ||
AK047691.1 | - | 4, 5 | 571 | |||
AK088218.1 | - | 4, 5 | 572 | |||
AK141504.1 | - | 1, 2 | 1350 | |||
AK142228.1 | - | 4, 5 | 543 | |||
BC096540.1 | - | 4, 5 | 526 | 6403681 |