Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178909.4 | NP_849240.1 | 8 | 953 | ||
GenBank mRNA | AK033294.1 | - | 8 | 899 | ||
AK047691.1 | - | 8 | 927 | |||
AK088218.1 | - | 8 | 928 | |||
AK142228.1 | - | 8 | 900 | |||
AK214428.1 | - | 3 | 236 | |||
BC096540.1 | - | 8 | 882 | 6403681 |