Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025936.3 | NP_080212.2 | 6 | 693 | ||
GenBank mRNA | AK011383.1 | - | 6 | 671 | ||
AK076160.1 | - | 6 | 668 | |||
AK149856.1 | - | 6 | 668 | |||
AK151966.1 | - | 5 | 491 | |||
AK152108.1 | - | 6 | 668 | |||
AK160216.1 | - | 6 | 668 | |||
AK166097.1 | - | 6 | 668 | |||
AK168194.1 | - | 6 | 668 | |||
BC020132.1 | - | 6 | 651 | 3989952 |