Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003941.2 | NP_001003941.1 | 8 | 1108 | ||
NM_001165036.1 | NP_001158508.1 | 8 | 1096 | |||
NM_002541.3 | NP_002532.2 | 8 | 1108 | |||
XM_005249759.4 | XP_005249816.1 | 9 | 1085 | |||
XM_005249761.3 | XP_005249818.1 | 9 | 1091 | |||
XM_011515408.2 | XP_011513710.1 | 10 | 1172 | |||
GenBank mRNA | AK294951.1 | - | 5 | 482 | ||
AK296400.1 | - | 5 | 609 | |||
AK302805.1 | - | 6 | 720 | |||
AK304439.1 | - | 8 | 1027 | |||
AK304690.1 | - | 6 | 691 | |||
AK314179.1 | - | 8 | 1030 | |||
BC004964.1 | - | 7 | 991 | 3528538 | ||
BC009580.1 | - | 8 | 1042 | |||
BC014617.2 | - | 7 | 991 | 3010095 | ||
BC128549.1 | - | 7 | 655 | |||
BC128550.1 | - | 7 | 667 | |||
D10523.1 | - | 8 | 1006 |