Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyhip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145981.3 | NP_666093.1 | 2 | 419 | ||
GenBank mRNA | AK049900.1 | - | 2 | 420 | ||
BC030494.1 | - | 2 | 416 | |||
BC139360.1 | - | 3 | 402 | 8862382 |