Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253813.1 | NP_001240742.1 | 9 | 1090 | ||
NM_175344.4 | NP_780553.2 | 8 | 1027 | |||
GenBank mRNA | AK089300.1 | - | 8 | 940 | ||
AK159411.1 | - | 9 | 1041 | |||
BC060732.1 | - | 8 | 1027 | 6409332 |