Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081066.1 | NP_001074535.1 | 19 | 3362 | ||
GenBank mRNA | AK053114.1 | - | 9 | 1696 | ||
AK173046.1 | - | 19 | 3262 | |||
BC132389.1 | - | 19 | 3250 | 40130666 | ||
BC137791.1 | - | 19 | 3250 | 8860810 | ||
BC144912.1 | - | 19 | 3250 |