Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkdcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134117.2 | NP_598878.2 | 3, 4 | 1222 | ||
GenBank mRNA | AB381893.1 | - | 3, 4 | 1214 | ||
AB436780.1 | - | 3, 4 | 1203 | |||
BC022157.1 | - | 2, 3 | 369 | |||
BC086639.1 | - | 4, 5 | 1060 | 6828629 |