Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027179.1 | NP_081455.1 | 6 | 545 | ||
NM_134118.5 | NP_598879.1 | 7 | 590 | |||
GenBank mRNA | AK002384.1 | - | 7 | 560 | ||
AK010513.1 | - | 6 | 481 | |||
AK010984.1 | - | 7 | 564 | |||
AK013558.1 | - | 7 | 543 | |||
AK020708.1 | - | 6 | 539 | |||
AK171718.1 | - | 2 | 297 | |||
AK172506.1 | - | 2 | 275 | |||
AK189199.1 | - | 3 | 121 | |||
BC019984.1 | - | 7 | 576 | 4240045 | ||
BC094046.1 | - | 7 | 690 | 5064002 |