Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134133.2 | NP_598894.1 | 2 | 1710 | ||
GenBank mRNA | AF313412.1 | - | 2 | 1252 | ||
AK008053.1 | - | 1 | 14 | |||
AK143383.1 | - | 2 | 1714 | |||
AK155873.1 | - | 2 | 1356 | |||
AK168283.1 | - | 2 | 1533 | |||
AK171810.1 | - | 2 | 1368 | |||
BC012898.1 | - | 2 | 1218 | 4013357 |