Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160356.1 | NP_001153828.1 | 1 | 173 | ||
GenBank mRNA | AK131602.1 | - | 1 | 164 | ||
AK131892.1 | - | 1 | 137 | |||
AK132116.1 | - | 1 | 164 | |||
AY941793.1 | - | 1 | 164 | |||
BC029863.1 | - | 1 | 98 | 4460168 |