Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4kb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293715.1 | NP_001280644.1 | 10 | 2511 | ||
NM_001293716.1 | NP_001280645.1 | 8 | 1529 | |||
NM_175356.3 | NP_780565.2 | 9 | 2466 | |||
GenBank mRNA | AK028974.1 | - | 9 | 2451 | ||
AK030704.1 | - | 9 | 2226 | |||
AK048405.1 | - | 8 | 1322 | |||
AK053614.1 | - | 9 | 2461 | |||
AK157918.1 | - | 9 | 2258 | |||
AK172074.1 | - | 9 | 2459 | |||
BC059895.1 | - | 2 | 213 | |||
BC079846.1 | - | 9 | 2466 | 5707855 | ||
BC113176.1 | - | 10 | 2366 | 40060773 | ||
BC113781.1 | - | 8 | 1429 | 40060770 | ||
BC138457.1 | - | 9 | 2369 | 8861477 | ||
BC138458.1 | - | 9 | 2369 | 8861478 |