Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172100.1 | NP_001165571.1 | 5 | 569 | ||
NM_001172101.1 | NP_001165572.1 | 5 | 572 | |||
NM_145135.3 | NP_660117.2 | 5 | 720 | |||
GenBank mRNA | AF071546.1 | - | 4 | 440 | ||
AK133822.1 | - | 5 | 500 | |||
AK141351.1 | - | 5 | 545 | |||
AK145173.1 | - | 5 | 554 | |||
AK147059.1 | - | 5 | 506 | |||
BC010331.1 | - | 5 | 589 | 2654341 |