Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001796.4 | NP_001001796.1 | 6 | 705 | ||
GenBank mRNA | AK039633.1 | - | 6 | 706 | ||
BC132613.1 | - | 6 | 674 | 40130890 | ||
BC145917.1 | - | 6 | 674 | 40131220 | ||
EU670678.1 | - | 7 | 825 |