Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023223.2 | NP_075712.2 | 9 | 1502 | ||
GenBank mRNA | AB045313.1 | - | 9 | 1406 | ||
AF312208.1 | - | 9 | 1399 | |||
AK011131.1 | - | 7 | 889 | |||
AK011723.1 | - | 9 | 1501 | |||
AK075998.1 | - | 9 | 1450 | |||
AK076030.1 | - | 9 | 1454 | |||
AK083459.1 | - | 9 | 1331 | |||
AK160204.1 | - | 9 | 1514 | |||
AK168650.1 | - | 9 | 1454 | |||
BC003215.1 | - | 9 | 1439 | 3157842 |