Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc14a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171010.1 | NP_001164481.1 | 6 | 977 | ||
NM_001171011.1 | NP_001164482.1 | 5 | 712 | |||
NM_028122.4 | NP_082398.1 | 5 | 772 | |||
GenBank mRNA | AF448798.1 | - | 4 | 569 | ||
AJ420967.1 | - | 5 | 631 | |||
AK012066.1 | - | 5 | 622 | |||
AK041979.1 | - | 5 | 714 | |||
AK153891.1 | - | 6 | 882 | |||
BC058594.1 | - | 5 | 659 | 5364683 | ||
BC086673.1 | - | 5 | 668 | 30646772 | ||
BC100570.1 | - | 5 | 659 | 30044353 |