Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A18AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302862.1 | NP_001289791.1 | 3 | 839 | ||
NM_007105.3 | NP_009036.2 | 4 | 1234 | |||
XM_017017832.1 | XP_016873321.1 | 2 | 2272 | |||
XM_017017833.1 | XP_016873322.1 | 4 | 2655 | |||
XM_017017834.1 | XP_016873323.1 | 2 | 1936 | |||
XM_017017835.1 | XP_016873324.1 | 2 | 1715 | |||
XM_017017836.1 | XP_016873325.1 | 2 | 580 | |||
GenBank mRNA | AF035407.1 | - | 2 | 601 | ||
AF037066.1 | - | 4 | 1030 | |||
BC030237.1 | - | 3 | 395 | 4185944 |