Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025315.3 | NP_079591.1 | 4 | 352 | ||
GenBank mRNA | AK009742.1 | - | 4 | 353 | ||
AK012217.1 | - | 4 | 353 | |||
AK014428.1 | - | 4 | 346 | |||
AK019417.1 | - | 4 | 342 | |||
BC012286.1 | - | 4 | 343 | 3493056 | ||
BC096641.1 | - | 3 | 1302 |