Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170555.1 | NP_001164026.1 | 2 | 242 | ||
NM_001170556.1 | NP_001164027.1 | 2 | 239 | |||
NM_145401.2 | NP_663376.2 | 6 | 1321 | |||
GenBank mRNA | AK010703.1 | - | 2 | 181 | ||
AK032238.1 | - | 4 | 833 | |||
AK154296.1 | - | 6 | 1322 | |||
AK165834.1 | - | 2 | 181 | |||
AY348864.1 | - | 2 | 172 | |||
BC015283.1 | - | 6 | 1109 | 4238045 | ||
BQ572978.1 | - | 2 | 172 | |||
BU707674.1 | - | 2 | 142 | |||
BY128345.1 | - | 2 | 242 |