Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2af1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024187.4 | NP_077149.2 | 3 | 208 | ||
GenBank mRNA | AK008332.1 | - | 3 | 170 | ||
AK012334.1 | - | 3 | 168 | |||
AK012849.1 | - | 3 | 168 | |||
AK088979.1 | - | 3 | 178 | |||
AK150872.1 | - | 3 | 208 | |||
BC002184.1 | - | 3 | 147 | |||
BC069888.1 | - | 3 | 150 | |||
BC115479.1 | - | 3 | 170 | 40100558 | ||
BC115480.1 | - | 3 | 170 | 40100559 |