Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2af1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163769.1 | NP_001157241.1 | 2 | 160 | ||
NM_024187.4 | NP_077149.2 | 2 | 160 | |||
GenBank mRNA | AK008332.1 | - | 2 | 122 | ||
AK012334.1 | - | 2 | 120 | |||
AK012849.1 | - | 2 | 120 | |||
AK088979.1 | - | 2 | 130 | |||
AK150872.1 | - | 2 | 160 | |||
AV453783.1 | - | 2 | 142 | |||
BC002184.1 | - | 2 | 99 | |||
BC069888.1 | - | 2 | 102 | |||
BC115479.1 | - | 2 | 122 | 40100558 | ||
BC115480.1 | - | 2 | 122 | 40100559 |