Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003911.2 | NP_001003911.2 | 19 | 3666 | ||
GenBank mRNA | AY551090.1 | - | 19 | 3525 | ||
BC058991.1 | - | 16 | 2638 | |||
BC076575.1 | - | 18 | 3374 | |||
BC141173.1 | - | 19 | 3442 | 9055785 | ||
BC145491.1 | - | 19 | 3442 | 9054054 |