Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178648.2 | NP_848763.2 | 8 | 724 | ||
GenBank mRNA | AK011496.1 | - | 8 | 712 | ||
AK016964.1 | - | 7 | 596 | |||
AK132565.1 | - | 8 | 717 | |||
AK161397.1 | - | 8 | 1119 | |||
BC024492.1 | - | 8 | 718 | 4981720 | ||
D88447.1 | - | 8 | 707 |