Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319108.1 | NP_001306037.1 | 2 | 276 | ||
NM_020530.5 | NP_065391.1 | 2 | 95 | |||
GenBank mRNA | BC011589.1 | - | 2 | 92 | 4548943 | |
BM562358.1 | - | 2 | 288 | |||
CR456534.1 | - | 2 | 65 | |||
CR541703.1 | - | 2 | 51 | |||
CR981101.1 | - | 2 | 95 |