Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033226.4 | NP_001028398.1 | 2 | 432 | ||
NM_001285895.1 | NP_001272824.1 | 3 | 396 | |||
NM_001285896.1 | NP_001272825.1 | 3 | 409 | |||
NM_001285897.1 | NP_001272826.1 | 3 | 651 | |||
NM_001285898.1 | NP_001272827.1 | 3 | 661 | |||
GenBank mRNA | AK077094.1 | - | 3 | 501 | ||
AK139540.1 | - | 3 | 411 | |||
AK163350.1 | - | 2 | 433 | |||
BC044740.2 | - | 3 | 661 | |||
JN255799.1 | - | 3 | 396 |