Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Als2cr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175370.4 | NP_780579.1 | 5 | 394 | ||
GenBank mRNA | AK016678.1 | - | 5 | 552 | ||
AK077190.1 | - | 5 | 362 | |||
AK133373.1 | - | 392 | ||||
BC145039.1 | - | 4 | 303 | |||
BC145040.1 | - | 4 | 299 |