Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178651.3 | NP_848766.2 | 9 | 937 | ||
GenBank mRNA | AF263460.1 | - | 7 | 491 | ||
AK028885.1 | - | 9 | 904 | |||
AK035541.1 | - | 9 | 924 | |||
AK038559.1 | - | 8 | 843 | |||
AK082853.1 | - | 9 | 939 | |||
AK143277.1 | - | 9 | 938 | |||
AK149549.1 | - | 2 | 1088 | |||
AK182342.1 | - | 1 | 15 | |||
AY682914.1 | - | 10 | 930 | |||
BC023833.1 | - | 1 | 41 | |||
BC026565.1 | - | 5 | 355 | |||
BC027806.1 | - | 9 | 699 | |||
BC031705.1 | - | 6 | 451 | |||
BC055773.1 | - | 6 | 390 | |||
BC078440.1 | - | 7 | 514 | |||
BC141008.1 | - | 10 | 903 | 9055620 |